2-[1-Methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-5-sulphonyl chloride 97%
Fornitore: Apollo Scientific
|
Pericolo
|
PC3008-100MG
PC3008-250MG
APOSPC3008-100MGEA
29.2
EUR
APOSPC3008-100MG
APOSPC3008-250MG
2-[1-Methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-5-sulphonyl chloride 97%
2-[1-Methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-5-sulphonyl chloride
Formula:
C₉H₆ClF₃N₂O₂S₂ Peso molecolare: 330,74 g/mol Punto di fusione: 77…80 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00052283 CAS Number: 166964-31-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...