Stampa…

Methyl-2-amino-4-(trifluoromethyl)benzoate

Fornitore: Apollo Scientific

Synonyms: Methyl 2-amino-4-(trifluoromethyl)benzoate, 2-Amino-4-(trifluoromethyl)benzoic acid methyll ester

PC300713-1G PC300713-5G
APOSPC300713-1GEA 62.6 EUR
APOSPC300713-1G APOSPC300713-5G
Methyl-2-amino-4-(trifluoromethyl)benzoate
Methyl-2-amino-4-(trifluoromethyl)benzoate
Formula: C₉H₈F₃NO₂
Peso molecolare: 219,1605 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08543939
CAS Number: 61500-87-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR