Stampa…

1-[4-(Trifluoromethyl)-2-pyrimidinyl]tetrahydro-4(1H)-pyridinone

Fornitore: Apollo Scientific

Synonyms: 1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidin-4-one

PC300676-10G PC300676-1G PC300676-5G
APOSPC300676-10GEA 663 EUR
APOSPC300676-10G APOSPC300676-1G APOSPC300676-5G
1-[4-(Trifluoromethyl)-2-pyrimidinyl]tetrahydro-4(1H)-pyridinone
1-[4-(Trifluoromethyl)-2-pyrimidinyl]tetrahydro-4(1H)-pyridinone
Formula: C₁₀H₁₀F₃N₃O
Peso molecolare: 245,21 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09027162
CAS Number: 937604-44-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR