Stampa…

Ethyl-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate

Fornitore: Apollo Scientific

PC300546-1G PC300546-5G
APOSPC300546-1GEA 154 EUR
APOSPC300546-1G APOSPC300546-5G
Ethyl-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Ethyl-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formula: C₁₂H₉F₃O₂S
Peso molecolare: 274,26 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11100111
CAS Number: 1002355-89-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR