Stampa…

2-Fluoro-3-oxo-3-phenylpropanenitrile 95%

Fornitore: Apollo Scientific

PC28206-1G PC28206-5G
APOSPC28206-1GEA 299 EUR
APOSPC28206-1G APOSPC28206-5G
2-Fluoro-3-oxo-3-phenylpropanenitrile 95%
2-Fluoro-3-oxo-3-phenylpropanenitrile
Formula: C₉H₆FNO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18426513
CAS Number: 343271-85-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR