Stampa…

2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoic acid

Fornitore: Apollo Scientific

Synonyms: Fmoc-p-fluoro-DL-Phe-OH

PC27881-1G PC27881-5G
APOSPC27881-1GEA 256 EUR
APOSPC27881-1G APOSPC27881-5G
2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoic acid
2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoic acid
Formula: C₂₄H₂₀FNO₄
Peso molecolare: 405,425 g/mol
Temperatura di stoccaggio: Ambiente
CAS Number: 264276-42-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR