Stampa…
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
Fornitore: Apollo Scientific
Pericolo
Synonyms: 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
PC2519-25G PC2519-5G PC2519-100G
APOSPC2519-25GEA 21.9 EUR
APOSPC2519-25G APOSPC2519-5G APOSPC2519-100G
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
Peptide coupling reagent.
Formula: C₁₂H₂₂F₆N₆OP₂
Peso molecolare: 442,28 g/mol
Punto di fusione: 136…140 °C (dec.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00011948
CAS Number: 56602-33-6
EINECS: 260-279-1
UN: 1325
ADR: 4.1,II