Stampa…

3-Bromo-5-fluorobenzyl alcohol 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: (3-Bromo-5-fluorophenyl)methanol

PC2406-1G PC2406-25G PC2406-5G
APOSPC2406-1GEA 21.9 EUR
APOSPC2406-1G APOSPC2406-25G APOSPC2406-5G
3-Bromo-5-fluorobenzyl alcohol 98%
3-Bromo-5-fluorobenzyl alcohol
Formula: C₇H₆BrFO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04116320
CAS Number: 216755-56-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR