5-Chloro-3-(trifluoromethyl)-1H-pyrazole 95+%
Fornitore: Apollo Scientific
PC201118-100MG
PC201118-250MG
APOSPC201118-100MGEA
96
EUR
APOSPC201118-100MG
APOSPC201118-250MG
5-Chloro-3-(trifluoromethyl)-1H-pyrazole 95+%
5-Chloro-3-(trifluoromethyl)-1H-pyrazole
Formula:
C₄H₂ClF₃N₂ Peso molecolare: 170,52 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD07375186 CAS Number: 131797-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...