4-Amino-5-ethynyl-2-(trifluoromethyl)benzenecarbonitrile
Fornitore: Apollo Scientific
Formula:
C₁₀H₅F₃N₂ Punto di fusione: 122…123 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD20921572 CAS Number: 949513-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...