Stampa…

3-(Trifluoromethyl)phenylboronic acid pinacol ester

Fornitore: Apollo Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(trifluoromethyl)phenyl]-1,3,2-dioxaborolane, 3-(Trifluoromethyl)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)benzene

PC200204-1G PC200204-5G
APOSPC200204-1GEA 58.2 EUR
APOSPC200204-1G APOSPC200204-5G
3-(Trifluoromethyl)phenylboronic acid pinacol ester
3-(Trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₆BF₃O₂
Peso molecolare: 272,0712 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05863925
CAS Number: 325142-82-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR