2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
Fornitore: Apollo Scientific
|
Attenzione
|
PC200202-1G
PC200202-5G
APOSPC200202-1GEA
109
EUR
APOSPC200202-1G
APOSPC200202-5G
2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
Formula:
C₈H₄ClF₃N₂ Peso molecolare: 220,58 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD10568352 CAS Number: 13600-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...