Stampa…

Potassium-4-(1-piperidinylcarbonyl)phenyltrifluoroborate

Fornitore: Apollo Scientific

Synonyms: Potassium trifluoro[4-(piperidine-1-carbonyl)phenyl]boranuide

PC200141-10G PC200141-1G PC200141-5G
APOSPC200141-10GEA 346 EUR
APOSPC200141-10G APOSPC200141-1G APOSPC200141-5G
Potassium-4-(1-piperidinylcarbonyl)phenyltrifluoroborate
Potassium-4-(1-piperidinylcarbonyl)phenyltrifluoroborate
Formula: C₁₂H₁₄BF₃KNO
Peso molecolare: 295,15 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11977715
CAS Number: 1359865-98-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR