Stampa…

2-(4-Fluorophenyl)propanoic acid

Fornitore: Apollo Scientific
Pericolo

PC200089-1G PC200089-5G
APOSPC200089-1GEA 87.3 EUR
APOSPC200089-1G APOSPC200089-5G
2-(4-Fluorophenyl)propanoic acid
2-(4-Fluorophenyl)propanoic acid
Formula: C₉H₉FO₂
Peso molecolare: 168,1650 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00054492
CAS Number: 75908-73-5
EINECS: 278-340-6
UN: 2811
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR