Potassium-3-carboxy-5-nitrophenyltrifluoroborate 96%
Fornitore: Apollo Scientific
PC1696-100G
PC1696-25G
PC1696-5G
APOSPC1696-100GEA
786
EUR
APOSPC1696-100G
APOSPC1696-25G
APOSPC1696-5G
Potassium-3-carboxy-5-nitrophenyltrifluoroborate 96%
Potassium-3-carboxy-5-nitrophenyltrifluoroborate
Formula:
C₇H₄BF₃KNO₄ Punto di fusione: >300 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04115772 CAS Number: 850623-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...