Stampa…

Phenyl trifluoromethyl sulphoxide 98%

Fornitore: Apollo Scientific

Synonyms: ((Trifluoromethyl)sulphinyl)benzene, [(Trifluoromethyl)sulphinyl]benzene

PC10685-1G PC10685-5G
APOSPC10685-1GEA 125 EUR
APOSPC10685-1G APOSPC10685-5G
Phenyl trifluoromethyl sulphoxide 98%
Phenyl trifluoromethyl sulphoxide
Formula: C₇H₅F₃OS
Peso molecolare: 194,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD30475661
CAS Number: 703-18-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR