Stampa…

Silver(I) trifluoromethanethiolate 95%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Trifluoromethanethiol Silver(I) salt

PC10683-1G PC10683-5G
APOSPC10683-1GEA 179 EUR
APOSPC10683-1G APOSPC10683-5G
Silver(I) trifluoromethanethiolate 95%
Silver(I) trifluoromethanethiolate
Formula: CAgF₃S
Peso molecolare: 208,93 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD25563234
CAS Number: 811-68-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR