Stampa…

5-Bromo-2-fluoro-3-pyridineboronic acid

Fornitore: Apollo Scientific
Attenzione

Synonyms: 5-Bromo-2-fluoro-3-pyridylboronic, 5-Bromo-2-fluoro-3-pyridinylboronic acid, 5-Bromo-2-fluoropyridine-3-boronic acid

PC10510-100G PC10510-25G PC10510-5G
APOSPC10510-100GEA 326 EUR
APOSPC10510-100G APOSPC10510-25G APOSPC10510-5G
5-Bromo-2-fluoro-3-pyridineboronic acid
5-Bromo-2-fluoro-3-pyridineboronic acid
Formula: C₅H₄BBrFNO₂
Peso molecolare: 219,8 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04039317
CAS Number: 501435-91-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR