2-Amino-5-(3-fluorophenyl)-4-thiazolecarbohydrazide
Fornitore: Apollo Scientific
PC10365-1G
PC10365-500MG
PC10365-5G
APOSPC10365-1GEA
217
EUR
APOSPC10365-1G
APOSPC10365-500MG
APOSPC10365-5G
2-Amino-5-(3-fluorophenyl)-4-thiazolecarbohydrazide
2-Amino-5-(3-fluorophenyl)-4-thiazolecarbohydrazide
Formula:
C₁₀H₉FN₄OS Punto di fusione: 122 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD06200940 CAS Number: 886361-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...