Stampa…

Aluminium trifluoromethanesulphonate 99%

Fornitore: Apollo Scientific
Pericolo

Synonyms: Aluminum trifluoromethanesulphonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)

PC1010-10G PC1010-25G
APOSPC1010-10GEA 132 EUR
APOSPC1010-10G APOSPC1010-25G
Aluminium trifluoromethanesulphonate 99%
Aluminium trifluoromethanesulphonate
Formula: C₃AlF₉O₉S₃
Peso molecolare: 474,19 g/mol
Punto di fusione: 300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00143596
CAS Number: 74974-61-1
UN: 3261
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR