FCCP 95+%
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
FCCP, [4-(Trifluoromethoxy)phenyl]carbonohydrazonoyl dicyanide, [4-(Trifluoromethoxy)phenylhydrazono]malononitrile, Carbonyl cyanide 4-(Trifluoromethoxy)phenylhydrazone
Formula:
C₁₀H₅F₃N₄O Punto di fusione: 174…175 °C (dec.) Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00009699 CAS Number: 370-86-5 EINECS: 206-730-8 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...