Stampa…

3-Chloro-2,4-difluorobenzoyl chloride

Fornitore: Apollo Scientific
Pericolo

Synonyms: 3-Chloro-2,4-difluorobenzene-1-carbonyl chloride

PC0161-25G PC0161-5G
APOSPC0161-25GEA 541 EUR
APOSPC0161-25G APOSPC0161-5G
3-Chloro-2,4-difluorobenzoyl chloride
3-Chloro-2,4-difluorobenzoyl chloride
Formula: C₇H₂Cl₂F₂O
Temperatura di stoccaggio: Ambiente
CAS Number: 157373-00-7
UN: 1760
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR