6-Chloro-2-fluoro-3-methylbenzylamine
Fornitore: Apollo Scientific
|
Pericolo
|
PC0113-10G
PC0113-1G
PC0113-5G
PC0113-25G
APOSPC0113-10GEA
282
EUR
APOSPC0113-10G
APOSPC0113-1G
APOSPC0113-5G
APOSPC0113-25G
6-Chloro-2-fluoro-3-methylbenzylamine
6-Chloro-2-fluoro-3-methylbenzylamine
Formula:
C₈H₉ClFN Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01631420 CAS Number: 261762-86-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...