(Ethylsulphonyl)benzene
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
Ethyl phenyl sulphone, (Ethylsulphonyl)benzol
OR9713-100G
OR9713-25G
APOSOR9713-100GEA
72.7
EUR
APOSOR9713-100G
APOSOR9713-25G
(Ethylsulphonyl)benzene
(Ethylsulphonyl)benzene
Formula:
C₈H₁₀O₂S Peso molecolare: 170,23 g/mol Punto di ebollizione: 103 °C/0,75mmHg Punto di fusione: 35…44 °C Densità: 1,14 g/cm³ (25 °C) Flash Pt: 110 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00025081 CAS Number: 599-70-2 EINECS: 209-972-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...