Stampa…

6,7-Dihydrobenzo[b]thiophen-4-one 95%

Fornitore: Apollo Scientific

Synonyms: 4-Oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-Dihydro-4-benzo[b]thiophenone, 4-Keto-4,5,6,7-tetrahydrothianaphthene

OR965240-1G OR965240-5G
APOSOR965240-1GEA 61.1 EUR
APOSOR965240-1G APOSOR965240-5G
6,7-Dihydrobenzo[b]thiophen-4-one 95%
6,7-Dihydrobenzo[b]thiophen-4-one
Formula: C₈H₈OS
Peso molecolare: 152,211 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005861
CAS Number: 13414-95-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR