Stampa…

1-Chloro-7-methylisoquinoline 95%

Fornitore: Apollo Scientific

OR964809-100MG OR964809-1G OR964809-250MG
APOSOR964809-100MGEA 183 EUR
APOSOR964809-100MG APOSOR964809-1G APOSOR964809-250MG
1-Chloro-7-methylisoquinoline 95%
1-Chloro-7-methylisoquinoline
Formula: C₁₀H₈ClN
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09907915
CAS Number: 24188-80-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR