Stampa…

Methyl-N-acetyl-L-leucinate 95%

Fornitore: Apollo Scientific

Synonyms: Ac-Leu-OMe

OR964340-1G OR964340-25G OR964340-5G
APOSOR964340-1GEA 102 EUR
APOSOR964340-1G APOSOR964340-25G APOSOR964340-5G
Methyl-N-acetyl-L-leucinate 95%
Methyl-N-acetyl-L-leucinate
Formula: C₉H₁₇NO₃
Peso molecolare: 187,239 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00038292
CAS Number: 1492-11-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR