Boc-D-Lysinol(Z) 98%
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₁₉H₃₀N₂O₅ Temperatura di stoccaggio: Ambiente |
CAS Number:
252940-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...