2-Methyl-4-piperidone hydrochloride 95%
Fornitore: Apollo Scientific
Synonyms:
2-Methylpiperidin-4-one hydrochloride
OR964017-1G
OR964017-250MG
APOSOR964017-1GEA
173
EUR
APOSOR964017-1G
APOSOR964017-250MG
2-Methyl-4-piperidone hydrochloride 95%
2-Methyl-4-piperidone hydrochloride
Formula:
C₆H₁₂ClNO Peso molecolare: 149,62 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08669336 CAS Number: 13729-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...