Stampa…

Boc-N-methyl-D-Valine 97%

Fornitore: Apollo Scientific

Synonyms: Boc-D-N-Me-Val-OH, N-Boc-N-methyl-D-valine, (R)-2-((tert-Butoxycarbonyl)(methyl)amino)-3-methylbutanoic acid, Boc-N-Me-D-Val-OH

OR963882-1G OR963882-5G
APOSOR963882-1GEA 53.9 EUR
APOSOR963882-1G APOSOR963882-5G
Boc-N-methyl-D-Valine 97%
Boc-N-methyl-D-Valine
Formula: C₁₁H₂₁NO₄
Peso molecolare: 231,292 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00076996
CAS Number: 89536-85-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR