Stampa…

N-Methylpyridin-3-amine 98%

Fornitore: Apollo Scientific

Synonyms: 3-(Methylamino)pyridine

OR963110-10G OR963110-25G
APOSOR963110-10GEA 91.6 EUR
APOSOR963110-10G APOSOR963110-25G
N-Methylpyridin-3-amine 98%
N-Methylpyridin-3-amine
Formula: C₆H₈N₂
Peso molecolare: 108,144 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01418130
CAS Number: 18364-47-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR