Stampa…

3-(2-Methoxyphenoxy)propionic acid 95%

Fornitore: Apollo Scientific

OR962573-1G OR962573-5G
APOSOR962573-1GEA 142 EUR
APOSOR962573-1G APOSOR962573-5G
3-(2-Methoxyphenoxy)propionic acid 95%
3-(2-Methoxyphenoxy)propionic acid
Formula: C₁₀H₁₂O₄
Peso molecolare: 196,1999 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01931914
CAS Number: 20370-80-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR