Stampa…

2-Methylquinoline-4-carboxamide 95%

Fornitore: Apollo Scientific

OR962451-10G OR962451-1G OR962451-5G
APOSOR962451-10GEA 865 EUR
APOSOR962451-10G APOSOR962451-1G APOSOR962451-5G
2-Methylquinoline-4-carboxamide 95%
2-Methylquinoline-4-carboxamide
Formula: C₁₁H₁₀N₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00023982
CAS Number: 15821-13-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR