1-Boc-4-methylpiperidine 97%
Fornitore: Apollo Scientific
Synonyms:
tert-Butyl 4-methylpiperidine-1-carboxylate
OR961738-1G
OR961738-5G
APOSOR961738-1GEA
227
EUR
APOSOR961738-1G
APOSOR961738-5G
1-Boc-4-methylpiperidine 97%
1-Boc-4-methylpiperidine
Formula:
C₁₁H₂₁NO₂ Peso molecolare: 199,2899 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04114947 CAS Number: 123387-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...