Stampa…

3-(Methylthio)-2,4-triazine 97%

Fornitore: Apollo Scientific

Synonyms: 3-(Methylthio)-as-Triazine

OR961054-10G OR961054-1G OR961054-5G
APOSOR961054-10GEA 111 EUR
APOSOR961054-10G APOSOR961054-1G APOSOR961054-5G
3-(Methylthio)-2,4-triazine 97%
3-(Methylthio)-2,4-triazine
Formula: C₄H₅N₃S
Peso molecolare: 127,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00490024
CAS Number: 28735-21-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR