Stampa…

4-(Isopropylsulphonyl)aniline 98%

Fornitore: Apollo Scientific

Synonyms: 4-(Propane-2-sulphonyl)aniline

OR960092-1G OR960092-250MG
APOSOR960092-1GEA 539 EUR
APOSOR960092-1G APOSOR960092-250MG
4-(Isopropylsulphonyl)aniline 98%
4-(Isopropylsulphonyl)aniline
Formula: C₉H₁₃NO₂S
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01566617
CAS Number: 25355-76-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR