2,4-Dichloropyrimidine-5-carbaldehyde 98%
Fornitore: Apollo Scientific
OR959527-1G
OR959527-250MG
OR959527-5G
APOSOR959527-1GEA
111
EUR
APOSOR959527-1G
APOSOR959527-250MG
APOSOR959527-5G
2,4-Dichloropyrimidine-5-carbaldehyde 98%
2,4-Dichloropyrimidine-5-carbaldehyde
Formula:
C₅H₂Cl₂N₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11112096 CAS Number: 871254-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...