Stampa…

2-Methylisoquinolin-1(2H)-one 98%

Fornitore: Apollo Scientific

OR958031-1G OR958031-250MG OR958031-5G
APOSOR958031-1GEA 219 EUR
APOSOR958031-1G APOSOR958031-250MG APOSOR958031-5G
2-Methylisoquinolin-1(2H)-one 98%
2-Methylisoquinolin-1(2H)-one
Formula: C₁₀H₉NO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00040275
CAS Number: 4594-71-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR