Stampa…

4-Chloro-3,5-dimethyl-1H-pyrazole 95%

Fornitore: Apollo Scientific

OR956934-1G OR956934-25G OR956934-5G
APOSOR956934-1GEA 132 EUR
APOSOR956934-1G APOSOR956934-25G APOSOR956934-5G
4-Chloro-3,5-dimethyl-1H-pyrazole 95%
4-Chloro-3,5-dimethyl-1H-pyrazole
Formula: C₅H₇ClN₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00825711
CAS Number: 15953-73-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR