Stampa…

3-Oxopentanoic acid 98%

Fornitore: Apollo Scientific

OR956096-1G OR956096-5G
APOSOR956096-1GEA 133 EUR
APOSOR956096-1G APOSOR956096-5G
3-Oxopentanoic acid 98%
3-Oxopentanoic acid
Formula: C₅H₈O₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11040434
CAS Number: 10191-25-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR