Stampa…

Methyl-4-methylpicolinate 97%

Fornitore: Apollo Scientific

Synonyms: Methyl 4-methylpicolinate

OR955702-1G OR955702-5G
APOSOR955702-1GEA 90.2 EUR
APOSOR955702-1G APOSOR955702-5G
Methyl-4-methylpicolinate 97%
Methyl-4-methylpicolinate
Formula: C₈H₉NO₂
Peso molecolare: 151,1600 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09265118
CAS Number: 13509-13-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR