Stampa…

Methyl-2,2-dimethylphenylacetate 95%

Fornitore: Apollo Scientific

Synonyms: Methyl 2-methyl-2-phenylpropanoate

OR955592-1G OR955592-5G
APOSOR955592-1GEA 48.1 EUR
APOSOR955592-1G APOSOR955592-5G
Methyl-2,2-dimethylphenylacetate 95%
Methyl-2,2-dimethylphenylacetate
Formula: C₁₁H₁₄O₂
Peso molecolare: 178,231 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01846451
CAS Number: 57625-74-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR