Stampa…

Ethylene glycol monoallyl ether 95%

Fornitore: Apollo Scientific

Synonyms: 2-(Allyloxy)ethanol, 2-Allyloxyethanol, 2-(2-Propenyloxy)ethanol, Allyl glycol

OR955005-100G OR955005-1KG OR955005-500G
APOSOR955005-100GEA 125 EUR
APOSOR955005-100G APOSOR955005-1KG APOSOR955005-500G
Ethylene glycol monoallyl ether 95%
Ethylene glycol monoallyl ether
Formula: C₅H₁₀O₂
Peso molecolare: 102,13 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00020607
CAS Number: 111-45-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR