Stampa…

2-(2-Chlorophenyl)thioacetamide 98%

Fornitore: Apollo Scientific

Synonyms: 2-(2-Chlorophenyl)ethanethioamide

OR954558-10G OR954558-1G OR954558-5G
APOSOR954558-10GEA 757 EUR
APOSOR954558-10G APOSOR954558-1G APOSOR954558-5G
2-(2-Chlorophenyl)thioacetamide 98%
2-(2-Chlorophenyl)thioacetamide
Formula: C₈H₈ClNS
Peso molecolare: 185,67 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00278248
CAS Number: 673476-96-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR