1-Ethyl-1H-pyrazole-3-carboxylic acid 98%
Fornitore: Apollo Scientific
Synonyms:
1-Ethylpyrazole-3-carboxylic Acid
OR952629-10G
OR952629-1G
OR952629-5G
APOSOR952629-10GEA
345
EUR
APOSOR952629-10G
APOSOR952629-1G
APOSOR952629-5G
1-Ethyl-1H-pyrazole-3-carboxylic acid 98%
1-Ethyl-1H-pyrazole-3-carboxylic acid
Formula:
C₆H₈N₂O₂ Peso molecolare: 140,14 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02090832 CAS Number: 400755-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...