Stampa…

3-Chloro-5-nitropyridin-4-ol 95%

Fornitore: Apollo Scientific

OR952268-1G OR952268-250MG
APOSOR952268-1GEA 109 EUR
APOSOR952268-1G APOSOR952268-250MG
3-Chloro-5-nitropyridin-4-ol 95%
3-Chloro-5-nitropyridin-4-ol
Formula: C₅H₃ClN₂O₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11044290
CAS Number: 31872-64-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR