1-(3-Chlorophenyl)ethanamine 98%
Fornitore: Apollo Scientific
Synonyms:
1-(3-Chlorophenyl)ethan-1-amine
OR951752-10G
OR951752-1G
OR951752-5G
APOSOR951752-10GEA
865
EUR
APOSOR951752-10G
APOSOR951752-1G
APOSOR951752-5G
1-(3-Chlorophenyl)ethanamine 98%
1-(3-Chlorophenyl)ethanamine
Formula:
C₈H₁₀ClN Peso molecolare: 155,63 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD05215231 CAS Number: 24358-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...