2-(Methanesulphonylamino)benzonitrile 98%
Fornitore: Apollo Scientific
OR951327-1G
OR951327-25G
OR951327-5G
APOSOR951327-1GEA
147
EUR
APOSOR951327-1G
APOSOR951327-25G
APOSOR951327-5G
2-(Methanesulphonylamino)benzonitrile 98%
2-(Methanesulphonylamino)benzonitrile
Formula:
C₈H₈N₂O₂S Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00590384 CAS Number: 50790-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...