Stampa…

(2,3-Dichlorophenyl)acetic acid methyl ester 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Methyl 2,3-Dichlorophenylacetate, Methyl (2,3-dichlorophenyl)acetate

OR951268-25G OR951268-5G OR951268-100G
APOSOR951268-25GEA 360 EUR
APOSOR951268-25G APOSOR951268-5G APOSOR951268-100G
(2,3-Dichlorophenyl)acetic acid methyl ester 98%
(2,3-Dichlorophenyl)acetic acid methyl ester
Formula: (C₆H₃Cl₂)CH₂COOCH₃
Peso molecolare: 219,07 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04112743
CAS Number: 10328-87-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR