Stampa…

4,7-Dichloroquinazoline 97%

Fornitore: Apollo Scientific

OR951087-10G OR951087-1G OR951087-5G
APOSOR951087-10GEA 216 EUR
APOSOR951087-10G APOSOR951087-1G APOSOR951087-5G
4,7-Dichloroquinazoline 97%
4,7-Dichloroquinazoline
Formula: C₈H₄Cl₂N₂
Peso molecolare: 199,0368 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00023911
CAS Number: 2148-57-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR